3-Mercapto-3-methyl-1-hexanol 99%
Supplier: Apollo Scientific
Warning
Synonyms: 3-Methyl-3-sulphanylhexan-1-ol
OR70181-1G OR70181-5G OR70181-250MG
APOSOR70181-1GEA 209 EUR
APOSOR70181-1G APOSOR70181-5G APOSOR70181-250MG
3-Mercapto-3-methyl-1-hexanol 99%
3-Mercapto-3-methyl-1-hexanol
Formula: C₇H₁₆OS
MW: 148.27 g/mol
Storage Temperature: Ambient
CAS Number: 307964-23-4