1-(4-Methoxyphenyl)ethanol
Supplier: Apollo Scientific
Warning
Synonyms: 4-Methoxy-alpha-methylbenzyl alcohol
OR70187-100G OR70187-25G
APOSOR70187-100GEA 219 EUR
APOSOR70187-100G APOSOR70187-25G
1-(4-Methoxyphenyl)ethanol
1-(4-Methoxyphenyl)ethanol
Formula: C₉H₁₂O₂
MW: 152.19 g/mol
Boiling Pt: 94…96 °C/1mmHg
Density: 1.079 g/cm³ (25 °C)
Flash Pt: 74 °C
Storage Temperature: Ambient
MDL Number: MFCD00016857
CAS Number: 3319-15-1