5,6-Dihydro-2H-pyran-2-one 97%
Supplier: Apollo Scientific
Warning
OR70226-1G OR70226-5G
APOSOR70226-1GEA 43.5 EUR
APOSOR70226-1G APOSOR70226-5G
5,6-Dihydro-2H-pyran-2-one 97%
5,6-Dihydro-2H-pyran-2-one
Formula: C₅H₆O₂
MW: 98.1 g/mol
Boiling Pt: 103 °C/10mmHg
Density: 1.139 g/cm³ (25 °C)
Flash Pt: 46 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00010439
CAS Number: 3393-45-1
UN: 1224
ADR: 3 III