1-Cyclopropylethylamine hydrochloride
Supplier: Apollo Scientific
Warning
Synonyms: 1-Cyclopropylethanamine hydrochloride
OR7051-10G OR7051-1G OR7051-5G
APOSOR7051-10GEA 575 EUR
APOSOR7051-10G APOSOR7051-1G APOSOR7051-5G
1-Cyclopropylethylamine hydrochloride
1-Cyclopropylethylamine hydrochloride
Formula: C₅H₁₂ClN
MW: 121.61 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06685893
CAS Number: 42390-64-7