3-[1,2,4]Triazol-1-ylpropionic acid 98%
Supplier: Apollo Scientific
Warning
Synonyms: 3-(1H-1,2,4-Triazol-1-yl)propanoic acid
OR7108-1G OR7108-5G
APOSOR7108-1GEA 154 EUR
APOSOR7108-1G APOSOR7108-5G
3-[1,2,4]Triazol-1-ylpropionic acid 98%
3-[1,2,4]Triazol-1-ylpropionic acid
Formula: C₅H₇N₃O₂
MW: 141.13 g/mol
Melting Pt: 177…179 °C
Storage Temperature: Ambient
MDL Number: MFCD02932735
CAS Number: 76686-84-5
UN: 1325
ADR: 4.1,III