3-Bromo-p-toluidine 99%
Supplier: Apollo Scientific
Danger
Synonyms: 4-Amino-2-bromotoluene, 3-Bromo-4-methylaniline
OR7118-100G OR7118-250G OR7118-500G
APOSOR7118-100GEA 63.8 EUR
APOSOR7118-100G APOSOR7118-250G APOSOR7118-500G
3-Bromo-p-toluidine 99%
3-Bromo-p-toluidine
Formula: C₇H₈BrN
MW: 186.05 g/mol
Boiling Pt: 254…257 °C
Melting Pt: 25…30 °C
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00134176
CAS Number: 7745-91-7
EINECS: 231-807-8
UN: 2811
ADR: 6.1 III