Cyclopentanone-2-carbonitrile 97%
Supplier: Apollo Scientific
Warning
OR7124-1G OR7124-25G OR7124-5G
APOSOR7124-1GEA 21.9 EUR
APOSOR7124-1G APOSOR7124-25G APOSOR7124-5G
Cyclopentanone-2-carbonitrile 97%
Cyclopentanone-2-carbonitrile
Formula: C₆H₇NO
MW: 109.13 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00154838
CAS Number: 2941-29-9