Iodosobenzene diacetate, Technical Grade
Supplier: Apollo Scientific
Warning
Synonyms: Iodobenzene diacetate, (Diacetoxyiodo)benzene
OR7138-100G OR7138-25G OR7138-500G
APOSOR7138-100GEA 29 EUR
APOSOR7138-100G APOSOR7138-25G APOSOR7138-500G
Iodosobenzene diacetate, Technical Grade
Iodosobenzene diacetate
Formula: C₁₀H₁₁IO₄
MW: 322.1 g/mol
Melting Pt: 159…163 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00008692
CAS Number: 3240-34-4
EINECS: 221-808-1