4-Benzyloxy-3,5-dimethylphenylboronic acid
Supplier: Apollo Scientific
Warning
OR7156-1G OR7156-5G OR7156-100MG OR7156-25G
APOSOR7156-1GEA 21.9 EUR
APOSOR7156-1G APOSOR7156-5G APOSOR7156-100MG APOSOR7156-25G
4-Benzyloxy-3,5-dimethylphenylboronic acid
4-Benzyloxy-3,5-dimethylphenylboronic acid
Formula: C₁₅H₁₇BO₃
Melting Pt: 212…214 °C
Storage Temperature: Refrigerator
MDL Number: MFCD04039003
CAS Number: 333788-94-6