4-Chloro-2-methylphenylboronic acid 98%
Supplier: Apollo Scientific
Warning
Synonyms: 4-Chloro-2-methylbenzeneboronic acid
OR7181-100G OR7181-25G
APOSOR7181-100GEA 215 EUR
APOSOR7181-100G APOSOR7181-25G
4-Chloro-2-methylphenylboronic acid 98%
4-Chloro-2-methylphenylboronic acid
Formula: C₇H₈BClO₂
MW: 170.40 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02683107
CAS Number: 209919-30-2