4-Chloro-3-methylphenylboronic acid
Supplier: Apollo Scientific
Warning
Synonyms: 4-Chloro-3-methylbenzeneboronic acid
OR7182-1G OR7182-25G OR7182-5G
APOSOR7182-1GEA 21.9 EUR
APOSOR7182-1G APOSOR7182-25G APOSOR7182-5G
4-Chloro-3-methylphenylboronic acid
4-Chloro-3-methylphenylboronic acid
Formula: C₇H₈BClO₂
MW: 170.40 g/mol
Melting Pt: 238…242 °C
Storage Temperature: Refrigerator
MDL Number: MFCD02683104
CAS Number: 161950-10-3