2,3-Dichlorophenylboronic acid
Supplier: Apollo Scientific
Warning
OR7196-1G OR7196-25G OR7196-5G
APOSOR7196-1GEA 29 EUR
APOSOR7196-1G APOSOR7196-25G APOSOR7196-5G
2,3-Dichlorophenylboronic acid
2,3-Dichlorophenylboronic acid
Formula: C₆H₅BCl₂O₂
MW: 190.82 g/mol
Melting Pt: 124…126 °C
Storage Temperature: Freezer
MDL Number: MFCD01075703
CAS Number: 151169-74-3