4-Methoxy-2,6-dimethylphenylboronic acid
Supplier: Apollo Scientific
Warning
Synonyms: 2,6-Dimethyl-4-methoxyphenylboronic acid
OR7207-1G OR7207-5G
APOSOR7207-1GEA 78.3 EUR
APOSOR7207-1G APOSOR7207-5G
4-Methoxy-2,6-dimethylphenylboronic acid
4-Methoxy-2,6-dimethylphenylboronic acid
Formula: C₉H₁₃BO₃
MW: 180.01 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD03452755
CAS Number: 361543-99-9