1-tert-Butyl-4-iodobenzene
Supplier: Apollo Scientific
Warning
OR7218-100G OR7218-25G
APOSOR7218-100GEA 91.5 EUR
APOSOR7218-100G APOSOR7218-25G
1-tert-Butyl-4-iodobenzene
1-tert-Butyl-4-iodobenzene
Formula: C₁₀H₁₃I
Boiling Pt: 243 °C
Density: 1.468 g/cm³ (25 °C)
Flash Pt: 106 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00052339
CAS Number: 35779-04-5