4-(Heptyloxy)phenylboronic acid
Supplier: Apollo Scientific
Warning
Synonyms: 4-Heptyloxyphenylboronic acid, 4-n-Heptyloxybenzeneboronic acid, 4-Heptyloxybenzeneboronic acid
OR7221-1G OR7221-25G OR7221-5G
APOSOR7221-1GEA 21.9 EUR
APOSOR7221-1G APOSOR7221-25G APOSOR7221-5G
4-(Heptyloxy)phenylboronic acid
4-(Heptyloxy)phenylboronic acid
Formula: C₁₃H₂₁BO₃
Storage Temperature: Ambient
MDL Number: MFCD04039169
CAS Number: 136370-19-9