4-(Morpholino)phenylboronic acid
Supplier: Apollo Scientific
Warning
Synonyms: (4-Morpholinophenyl)boronic acid
OR7242-1G OR7242-250MG OR7242-5G
APOSOR7242-1GEA 24.8 EUR
APOSOR7242-1G APOSOR7242-250MG APOSOR7242-5G
4-(Morpholino)phenylboronic acid
4-(Morpholino)phenylboronic acid
Formula: C₁₀H₁₄BNO₃
MW: 207.0341 g/mol
Melting Pt: 126…127 °C
Storage Temperature: Ambient
MDL Number: MFCD03095169
CAS Number: 186498-02-2