8-Nitroquinoline
Supplier: Apollo Scientific
Warning
OR7258-25G OR7258-5G OR7258-1G
APOSOR7258-25GEA 87.3 EUR
APOSOR7258-25G APOSOR7258-5G APOSOR7258-1G
8-Nitroquinoline
8-Nitroquinoline
Formula: C₉H₆N₂O₂
MW: 174.16 g/mol
Boiling Pt: 323
Melting Pt: 89…91 °C
Storage Temperature: Ambient
MDL Number: MFCD00006806
CAS Number: 607-35-2
EINECS: 210-135-9