Pyridine-3-boronic acid 1,3-propanediol ester
Supplier: Apollo Scientific
Warning
Synonyms: 3-(1,3,2-Dioxaborinan-2-yl)pyridine
OR7260-1G OR7260-5G
APOSOR7260-1GEA 0 EUR
APOSOR7260-1G APOSOR7260-5G
Pyridine-3-boronic acid 1,3-propanediol ester
Pyridine-3-boronic acid 1,3-propanediol ester
Formula: C₈H₁₀BNO₂
MW: 162.98 g/mol
Melting Pt: 100…104 °C
Storage Temperature: Ambient
MDL Number: MFCD01075749
CAS Number: 131534-65-1