2,4,6-Trivinylboroxin pyridine complex 95%
Supplier: Apollo Scientific
Warning
Synonyms: Trivinylcyclotriboroxan - Pyridine Complex
OR7267-1G OR7267-5G
APOSOR7267-1GEA 109 EUR
APOSOR7267-1G APOSOR7267-5G
2,4,6-Trivinylboroxin pyridine complex 95%
2,4,6-Trivinylboroxin pyridine complex
Formula: C₁₁H₁₄B₃NO₃
MW: 240.67 g/mol
Boiling Pt: 78…80 °C
Melting Pt: 44…52 °C
Flash Pt: 80 °C
Storage Temperature: Freezer
MDL Number: MFCD03839440
CAS Number: 95010-17-6