5-Isobutyl-1,3-cyclohexanedione 98%
Supplier: Apollo Scientific
Warning
OR7273-1G OR7273-5G
APOSOR7273-1GEA 33.5 EUR
APOSOR7273-1G APOSOR7273-5G
5-Isobutyl-1,3-cyclohexanedione 98%
5-Isobutyl-1,3-cyclohexanedione
Formula: C₁₀H₁₆O₂
Melting Pt: 125…129 °C
Storage Temperature: Ambient
MDL Number: MFCD00129458
CAS Number: 57641-95-9