5-Amino-3-methylisoxazole
Supplier: Apollo Scientific
Warning
Synonyms: 3-Methylisoxazol-5-ylamine, 3-Methylisoxazol-5-amine
OR7410-10G OR7410-1G OR7410-5G
APOSOR7410-10GEA 63.8 EUR
APOSOR7410-10G APOSOR7410-1G APOSOR7410-5G
5-Amino-3-methylisoxazole
5-Amino-3-methylisoxazole
Formula: C₄H₆N₂O
MW: 98.1 g/mol
Melting Pt: 81…83 °C
Storage Temperature: Ambient
MDL Number: MFCD00003151
CAS Number: 14678-02-5
EINECS: 238-719-9