Methyl-5-methoxy-2-nitrobenzoate 98%
Supplier: Apollo Scientific
Warning
Synonyms: Methyl 5-methoxy-2-nitrobenzoate, 5-Methoxy-2-nitrobenzoic acid methyl ester
OR7411-1G OR7411-25G OR7411-5G OR7411-100G
APOSOR7411-1GEA 14.7 EUR
APOSOR7411-1G APOSOR7411-25G APOSOR7411-5G APOSOR7411-100G
Methyl-5-methoxy-2-nitrobenzoate 98%
Methyl 5-methoxy-2-nitrobenzoate
Formula: C₉H₉NO₅
Melting Pt: 54…57 °C
Storage Temperature: Ambient
MDL Number: MFCD00662902
CAS Number: 2327-45-9