4-Bromo-2,5-xylenol 98%
Supplier: Apollo Scientific
Warning
Synonyms: 4-Bromo-2,5-dimethylphenol
OR7417-5G
APOSOR7417-5GEA 116 EUR
APOSOR7417-5G
4-Bromo-2,5-xylenol 98%
4-Bromo-2,5-xylenol
Formula: C₈H₉BrO
MW: 201.0600 g/mol
Melting Pt: 86…89 °C
Storage Temperature: Ambient
MDL Number: MFCD00029746
CAS Number: 85223-93-4