1-Acetyl-5-amino-2,3-dihydro-(1H)-indole, Technical Grade
Supplier: Apollo Scientific
Warning
Synonyms: 1-Acetyl-5-aminoindoline
OR7441-1G OR7441-250MG
APOSOR7441-1GEA 68.3 EUR
APOSOR7441-1G APOSOR7441-250MG
1-Acetyl-5-amino-2,3-dihydro-(1H)-indole, Technical Grade
1-Acetyl-5-amino-2,3-dihydro-(1H)-indole
Formula: C₁₀H₁₂N₂O
MW: 176.22 g/mol
Melting Pt: 181…184 °C
Storage Temperature: Ambient
MDL Number: MFCD00087208
CAS Number: 4993-96-8