4'-Methoxy[1,1'-biphenyl]-3-amine
Supplier: Apollo Scientific
Warning
Synonyms: 3-Amino-4'-methoxybiphenyl hydrochloride, 4'-Methoxy[1,1'-biphenyl]-3-amine
OR7448-1G OR7448-250MG
APOSOR7448-1GEA 231 EUR
APOSOR7448-1G APOSOR7448-250MG
4'-Methoxy[1,1'-biphenyl]-3-amine
4'-Methoxy[1,1'-biphenyl]-3-amine
Formula: C₁₃H₁₃NO
MW: 199.25 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03424668
CAS Number: 53059-28-2