3'-Methoxybiphenyl-4-carboxaldehyde
Supplier: Apollo Scientific
Warning
Synonyms: 3'-Methoxy-[1,1'-biphenyl]-4-carbaldehyde
OR7457-1G OR7457-250MG OR7457-5G
APOSOR7457-1GEA 109 EUR
APOSOR7457-1G APOSOR7457-250MG APOSOR7457-5G
3'-Methoxybiphenyl-4-carboxaldehyde
3'-Methoxybiphenyl-4-carboxaldehyde
Formula: C₁₄H₁₂O₂
MW: 212.248 g/mol
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD03424619
CAS Number: 209863-09-2