4'-Methylbiphenyl-2-ylamine hydrochloride
Supplier: Apollo Scientific
Warning
Synonyms: 2-Amino-4'-methylbiphenyl hydrochloride
OR7492-100MG OR7492-500MG
APOSOR7492-100MGEA 164 EUR
APOSOR7492-100MG APOSOR7492-500MG
4'-Methylbiphenyl-2-ylamine hydrochloride
4'-Methylbiphenyl-2-ylamine hydrochloride
Formula: C₁₃H₁₄ClN
MW: 219.72 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06656008
CAS Number: 1204-43-9