3-(Dimethylamino)acrylonitrile 97+%
Supplier: Apollo Scientific
Warning
OR7503-25G OR7503-1G OR7503-5G
APOSOR7503-25GEA 288 EUR
APOSOR7503-25G APOSOR7503-1G APOSOR7503-5G
3-(Dimethylamino)acrylonitrile 97+%
3-(Dimethylamino)acrylonitrile
Formula: C₅H₈N₂
MW: 96.13 g/mol
Boiling Pt: 76…80 °C/0.3mmHg
Density: 0.95 g/cm³ (25 °C)
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00001929
CAS Number: 2407-68-3
EINECS: 219-305-7
UN: 3276
ADR: 6.1,III