6-Bromo-1-hexene 96%
Supplier: Apollo Scientific
Warning
OR7533-100G OR7533-25G
APOSOR7533-100GEA 75.4 EUR
APOSOR7533-100G APOSOR7533-25G
6-Bromo-1-hexene 96%
6-Bromo-1-hexene
Formula: C₆H₁₁Br
MW: 163.06 g/mol
Boiling Pt: 47…51 °C/16mmHg
Density: 1.217 g/cm³ (25 °C)
Flash Pt: 54 °C
Storage Temperature: Ambient
MDL Number: MFCD00000269
CAS Number: 2695-47-8
EINECS: 220-267-9
UN: 1993
ADR: 3,III