1-Bromononane 98%
Supplier: Apollo Scientific
Warning
Synonyms: 1-Nonyl bromide
OR7540-25G OR7540-100G OR7540-500G
APOSOR7540-25GEA 89.9 EUR
APOSOR7540-25G APOSOR7540-100G APOSOR7540-500G
1-Bromononane 98%
1-Bromononane
Formula: BrH₂C(CH₂)₇CH₃
MW: 207.15 g/mol
Boiling Pt: 200…201 °C
Density: 1.084 g/cm³ (25 °C)
Flash Pt: 90 °C
Storage Temperature: Ambient
MDL Number: MFCD00000278
CAS Number: 693-58-3
EINECS: 211-755-2