2-Bromo-3-methoxypyridine
Supplier: Apollo Scientific
Danger
OR7550-10G OR7550-1G OR7550-5G
APOSOR7550-10GEA 60.9 EUR
APOSOR7550-10G APOSOR7550-1G APOSOR7550-5G
2-Bromo-3-methoxypyridine
2-Bromo-3-methoxypyridine
Formula: C₆H₆BrNO
MW: 188.02 g/mol
Melting Pt: 45…49 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD01570896
CAS Number: 24100-18-3
EINECS: 246-017-9