2-[4-(5-Bromopyrimidin-2-yl)piperazin-1-yl]ethanol 95+%
Supplier: Apollo Scientific
Warning
OR7608-1G OR7608-250MG
APOSOR7608-1GEA 365 EUR
APOSOR7608-1G APOSOR7608-250MG
2-[4-(5-Bromopyrimidin-2-yl)piperazin-1-yl]ethanol 95+%
2-[4-(5-Bromopyrimidin-2-yl)piperazin-1-yl]ethanol
Formula: C₁₀H₁₅BrN₄O
MW: 287.17 g/mol
Melting Pt: 97…99 °C
Storage Temperature: Ambient
MDL Number: MFCD03452782
CAS Number: 849021-42-7