6-tert-Butoxypyridine-2-carboxaldehyde 95+%
Supplier: Apollo Scientific
Warning
OR7622-1G OR7622-5G OR7622-10G
APOSOR7622-1GEA 365 EUR
APOSOR7622-1G APOSOR7622-5G APOSOR7622-10G
6-tert-Butoxypyridine-2-carboxaldehyde 95+%
6-tert-Butoxypyridine-2-carboxaldehyde
Formula: C₁₀H₁₃NO₂
MW: 179.22 g/mol
Boiling Pt: 46 °C/0.11mmHg
Storage Temperature: Ambient
MDL Number: MFCD03426051
CAS Number: 195044-13-4