2-Chloro-m-xylene 99%
Supplier: Apollo Scientific
Warning
Synonyms: 2-Chloro-1,3-dimethylbenzene, 1-Chloro-2,6-dimethylbenzene
OR7643-100G OR7643-25G OR7643-500G
APOSOR7643-100GEA 219 EUR
APOSOR7643-100G APOSOR7643-25G APOSOR7643-500G
2-Chloro-m-xylene 99%
2-Chloro-m-xylene
Formula: C₈H₉Cl
MW: 140.61 g/mol
Boiling Pt: 182…183 °C
Density: 1.053 g/cm³ (25 °C)
Flash Pt: 63 °C
Storage Temperature: Ambient
MDL Number: MFCD00000565
CAS Number: 6781-98-2
EINECS: 229-843-4