2-(4-Nitrophenyl)malondialdehyde 95+%
Supplier: Apollo Scientific
|
Warning
|
OR7644-1G
OR7644-5G
OR7644-25G
APOSOR7644-1GEA
102
EUR
APOSOR7644-1G
APOSOR7644-5G
APOSOR7644-25G
2-(4-Nitrophenyl)malondialdehyde 95+%
2-(4-Nitrophenyl)malondialdehyde
Formula:
C₉H₇NO₄ MW: 193.16 g/mol Melting Pt: 231…233 °C Storage Temperature: Ambient |
MDL Number:
MFCD02261926 CAS Number: 18915-53-2 |