1-Chloro-4-ethynylbenzene 98%
Supplier: Apollo Scientific
Danger
Synonyms: (4-Chlorophenyl)acetylene, 1-Ethynyl-4-chlorobenzene, (4-Chlorophenyl)ethyne
OR7651T-25G OR7651T-5G OR7651T-100G
APOSOR7651T-25GEA 84.1 EUR
APOSOR7651T-25G APOSOR7651T-5G APOSOR7651T-100G
1-Chloro-4-ethynylbenzene 98%
1-Chloro-4-ethynylbenzene
Formula: C₈H₅Cl
Melting Pt: 43…44 °C
Flash Pt: 10 °C
Storage Temperature: Ambient
MDL Number: MFCD00191917
CAS Number: 873-73-4
UN: 1325
ADR: 4.1,II