Pyrimidine-5-carboxaldehyde
Supplier: Apollo Scientific
Warning
Synonyms: 5-Pyrimidinecarboxaldehyde
OR7659-1G OR7659-5G
APOSOR7659-1GEA 29 EUR
APOSOR7659-1G APOSOR7659-5G
Pyrimidine-5-carboxaldehyde
Pyrimidine-5-carboxaldehyde
Formula: C₅H₄N₂O
MW: 108.1 g/mol
Boiling Pt: 80…82 °C/4mmHg
Melting Pt: 39…43 °C
Density: 1.23 g/cm³ (25 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD03426065
CAS Number: 10070-92-5