N-[3-(Aminomethyl)phenyl]-N-methylamine 97%
Supplier: Apollo Scientific
Danger
Synonyms: 3-(Methylamino)benzylamine, 3-(Aminomethyl)-N-methylaniline
OR7679-1G OR7679-5G
APOSOR7679-1GEA 74.1 EUR
APOSOR7679-1G APOSOR7679-5G
N-[3-(Aminomethyl)phenyl]-N-methylamine 97%
N-[3-(Aminomethyl)phenyl]-N-methylamine
Formula: C₈H₁₂N₂
Boiling Pt: 148…152 °C
Storage Temperature: Ambient
MDL Number: MFCD04037894
CAS Number: 768343-60-8
UN: 2735
ADR: 8,III