5-[5-(Piperidin-4-yl)-1,2,4-oxadiazol-3-yl]-1,3-benzodioxole 95+%
Supplier: Apollo Scientific
Warning
Synonyms: 4-[3-(1,3-Benzodioxol-5-yl)-1,2,4-oxadiazol-5-yl]piperidine
OR7684-1G OR7684-5G OR7684-25G
APOSOR7684-1GEA 246 EUR
APOSOR7684-1G APOSOR7684-5G APOSOR7684-25G
5-[5-(Piperidin-4-yl)-1,2,4-oxadiazol-3-yl]-1,3-benzodioxole 95+%
5-[5-(Piperidin-4-yl)-1,2,4-oxadiazol-3-yl]-1,3-benzodioxole
Formula: C₁₄H₁₅N₃O₃
MW: 273.29 g/mol
Melting Pt: 93…96 °C
Storage Temperature: Ambient
MDL Number: MFCD03426084
CAS Number: 849925-04-8