3-Methylbenzo[b]thiophene-2-carboxaldehyde 97%
Supplier: Apollo Scientific
OR7692-10G OR7692-25G OR7692-5G
APOSOR7692-10GEA 408 EUR
APOSOR7692-10G APOSOR7692-25G APOSOR7692-5G
3-Methylbenzo[b]thiophene-2-carboxaldehyde 97%
3-Methylbenzo[b]thiophene-2-carboxaldehyde
Formula: C₁₀H₈OS
Melting Pt: 88…90 °C
Storage Temperature: Ambient
MDL Number: MFCD00052307
CAS Number: 22053-74-3