Benzo[1,2,5]oxadiazol-4-ylamine
Supplier: Apollo Scientific
Warning
Synonyms: 4-Aminobenzofurazan
OR7721-1G OR7721-10G OR7721-5G
APOSOR7721-1GEA 49.3 EUR
APOSOR7721-1G APOSOR7721-10G APOSOR7721-5G
Benzo[1,2,5]oxadiazol-4-ylamine
Benzo[1,2,5]oxadiazol-4-ylamine
Formula: C₆H₅N₃O
MW: 135.13 g/mol
Melting Pt: 108…111 °C
Storage Temperature: Ambient
MDL Number: MFCD00168448
CAS Number: 767-63-5