N-(4-Aminophenyl)-N-ethyl-4,6-dimethyl-2-pyrimidinamine 95+%
Supplier: Apollo Scientific
Warning
Synonyms: N1-(4,6-Dimethylpyrimidin-2-yl)-N1-ethylbenzene-1,4-diamine
OR7729-1G OR7729-5G OR7729-25G
APOSOR7729-1GEA 185 EUR
APOSOR7729-1G APOSOR7729-5G APOSOR7729-25G
N-(4-Aminophenyl)-N-ethyl-4,6-dimethyl-2-pyrimidinamine 95+%
N-(4-Aminophenyl)-N-ethyl-4,6-dimethyl-2-pyrimidinamine
Formula: C₁₄H₁₈N₄
MW: 242.33 g/mol
Melting Pt: 135…138 °C
Storage Temperature: Ambient
MDL Number: MFCD00202979
CAS Number: 387358-43-2