4-Carboxyphenylboronic acid
Supplier: Apollo Scientific
Warning
OR7732-100G OR7732-25G OR7732-5G
APOSOR7732-100GEA 92.8 EUR
APOSOR7732-100G APOSOR7732-25G APOSOR7732-5G
4-Carboxyphenylboronic acid
4-Carboxyphenylboronic acid
Formula: C₇H₇BO₄
Melting Pt: 220 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00151801
CAS Number: 14047-29-1