3-Nitro-p-toluidine
Supplier: Apollo Scientific
Danger
Synonyms: 3-Nitro-4-methylaniline, 4-Methyl-3-nitroaniline, 4-Amino-2-nitrotoluene
OR7747-100G OR7747-25G OR7747-5G
APOSOR7747-100GEA 48 EUR
APOSOR7747-100G APOSOR7747-25G APOSOR7747-5G
3-Nitro-p-toluidine
3-Nitro-p-toluidine
Formula: C₇H₈N₂O₂
MW: 152.15 g/mol
Boiling Pt: 306
Melting Pt: 74…79 °C
Density: 1.312 g/cm³ (25 °C)
Flash Pt: 175 °C
Storage Temperature: Ambient
MDL Number: MFCD00007910
CAS Number: 119-32-4
EINECS: 204-314-0
UN: 2660
ADR: 6.1 III