N-[3-(Aminomethyl)phenyl]-N-methylacetamide hydrochloride 95+%
Supplier: Apollo Scientific
Warning
OR7776-1G OR7776-5G
APOSOR7776-1GEA 203 EUR
APOSOR7776-1G APOSOR7776-5G
N-[3-(Aminomethyl)phenyl]-N-methylacetamide hydrochloride 95+%
N-[3-(Aminomethyl)phenyl]-N-methylacetamide hydrochloride
Formula: C₁₀H₁₅ClN₂O
Melting Pt: 188…191 °C
Storage Temperature: Ambient
MDL Number: MFCD04039172
CAS Number: 849020-90-2