4-{3-[4-(Methylsulphonyl)phenyl]-1,2,4-oxadiazol-5-yl}piperidine 95+%
Supplier: Apollo Scientific
Warning
OR7798-1G OR7798-250MG
APOSOR7798-1GEA 92.8 EUR
APOSOR7798-1G APOSOR7798-250MG
4-{3-[4-(Methylsulphonyl)phenyl]-1,2,4-oxadiazol-5-yl}piperidine 95+%
4-{3-[4-(Methylsulphonyl)phenyl]-1,2,4-oxadiazol-5-yl}piperidine
Formula: C₁₄H₁₇N₃O₃S
Melting Pt: 128…131 °C
Storage Temperature: Ambient
MDL Number: MFCD03426082
CAS Number: 849925-03-7