5-(Methylsulphonyl)-2-piperidin-1-ylaniline 95+%
Supplier: Apollo Scientific
Warning
OR7804-1G OR7804-5G OR7804-10G
APOSOR7804-1GEA 285 EUR
APOSOR7804-1G APOSOR7804-5G APOSOR7804-10G
5-(Methylsulphonyl)-2-piperidin-1-ylaniline 95+%
5-(Methylsulphonyl)-2-piperidin-1-ylaniline
Formula: C₁₂H₁₈N₂O₂S
MW: 254.35 g/mol
Melting Pt: 128…129 °C
Storage Temperature: Refrigerator
MDL Number: MFCD04037964
CAS Number: 849035-90-1