4-[5-(Piperidin-4-yl)[1,2,4]oxadiazol-3-yl]pyridine 95+%
Supplier: Apollo Scientific
Warning
Synonyms: 5-(Piperidin-4-yl)-3-(pyridin-4-yl)-1,2,4-oxadiazole
OR7809-1G OR7809-5G
APOSOR7809-1GEA 270 EUR
APOSOR7809-1G APOSOR7809-5G
4-[5-(Piperidin-4-yl)[1,2,4]oxadiazol-3-yl]pyridine 95+%
4-[5-(Piperidin-4-yl)[1,2,4]oxadiazol-3-yl]pyridine
Formula: C₁₂H₁₄N₄O
MW: 230.27 g/mol
Melting Pt: 78…80 °C
Storage Temperature: Ambient
MDL Number: MFCD03426075
CAS Number: 276237-03-7