1-(2,5-Dichlorophenyl)biguanide hydrochloride
Supplier: Apollo Scientific
Warning
OR7819-100G OR7819-25G OR7819-5G
APOSOR7819-100GEA 882 EUR
APOSOR7819-100G APOSOR7819-25G APOSOR7819-5G
1-(2,5-Dichlorophenyl)biguanide hydrochloride
1-(2,5-Dichlorophenyl)biguanide hydrochloride
Formula: C₈H₁₀Cl₃N₅
Melting Pt: 211…215 °C
Storage Temperature: Ambient
MDL Number: MFCD00053018
CAS Number: 4767-32-2