3,6-Dimethoxypyridazine-4-carboxylic acid 96%
Supplier: Apollo Scientific
Warning
OR7843-1G OR7843-5G
APOSOR7843-1GEA 161 EUR
APOSOR7843-1G APOSOR7843-5G
3,6-Dimethoxypyridazine-4-carboxylic acid 96%
3,6-Dimethoxypyridazine-4-carboxylic acid
Formula: C₇H₈N₂O₄
MW: 184.15 g/mol
Melting Pt: 150…152 °C
Storage Temperature: Ambient
MDL Number: MFCD03426073
CAS Number: 89694-24-6